Assay Details
| Gene Symbol: Pragmin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107315.1 | NP_001100785.1 | 3 - 4 | 2336 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pragmin| Gene Symbol: Pragmin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107315.1 | NP_001100785.1 | 3 - 4 | 2336 | 104 | |
Set Membership: |
|