Assay Details
| Gene Symbol: Pragmin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107315.1 | NP_001100785.1 | 5 - 6 | 3250 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P165616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24402 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P165616 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P165616 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P165616 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P384506 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78010 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10791994 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791993 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791991 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791990 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791992 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10791989 | - | - |
| Rat Genome 230 2.0 Array | 1391121_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391121_at | - | - |
| Affymetrix HT Rat Focus Array | 1391121_at | - | - |
| Affymetrix HT Rat Focus Array | 1391121_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600012974.rn.1 | TC1600001746.rn.1 | RTA TC |
| Rat Expression Set 230 | 1391121_at | - | - |
| Rat Expression Set 230 | 1391121_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791989 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650690 | - | - |

Pragmin
Genomic Map