Assay Details
| Gene Symbol: Defal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033073.1 | NP_001028245.1 | 1 - 2 | 168 | 70 | |
| GenBank mRNA | AY623756.1 | - | 1 - 2 | 168 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Defal1
Genomic Map