Assay Details
Gene Symbol: RGD1304810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107314.1 | NP_001100784.1 | 5 - 6 | 1082 | 89 | |
XM_006253222.1 | XP_006253284.1 | 5 - 6 | 914 | 89 | ||
XM_006253223.2 | XP_006253285.1 | 5 - 6 | 2656 | 89 | ||
XM_008771300.1 | XP_008769522.1 | 4 - 5 | 1013 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P116028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131976 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P116028 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P116028 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P116028 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P511042 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95252 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10791986 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791985 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791984 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10791983 | - | - |
Rat Genome 230 2.0 Array | 1393353_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393353_at | - | - |
Affymetrix HT Rat Focus Array | 1393353_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393353_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600012949.rn.1 | TC1600001739.rn.1 | RTA TC |
Rat Expression Set 230 | 1393353_PM_at | - | - |
Rat Expression Set 230 | 1393353_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791983 | - | - |
Rat Genome U34 Set | rc_AI044301_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21311418 | - | - |
ABI Rat Genome Survey Microarray | 21974747 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040732 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369350 | - | - |
