Assay Details
| Gene Symbol: RGD1560672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109073.1 | NP_001102543.1 | 1 - 2 | 234 | 78 | |
| XM_006250221.2 | XP_006250283.1 | 1 - 2 | 447 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109073.1, XM_006250221.2 transcripts.
Set Membership: |
|

RGD1560672
Genomic Map