Assay Details
| Gene Symbol: Rxrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031765.1 | NP_113953.1 | 7 - 8 | 1279 | 64 | |
| XM_006250212.2 | XP_006250274.1 | 6 - 7 | 1423 | 64 | ||
| GenBank mRNA | AJ223083.1 | - | 6 - 7 | 814 | 64 | |
| BC091363.1 | - | 7 - 8 | 1279 | 7319081 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rxrg
Genomic Map