Assay Details
| Gene Symbol: Brinp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173121.1 | NP_775144.1 | 4 - 5 | 1059 | 83 | |
| XM_006249965.2 | XP_006250027.1 | 4 - 5 | 1477 | 83 | ||
| GenBank mRNA | AB077854.1 | - | 4 - 5 | 1059 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brinp3
Genomic Map