Assay Details
| Gene Symbol: Tnfsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053552.1 | NP_446004.1 | 1 - 2 | 327 | 63 | |
| GenBank mRNA | AF037067.1 | - | 1 - 2 | 327 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnfsf4
Genomic Map