Assay Details
| Gene Symbol: Tor1aip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145092.2 | NP_659560.2 | 8 - 9 | 1099 | 84 | |
| XM_006250027.2 | XP_006250089.1 | 8 - 9 | 644 | 84 | ||
| XM_006250028.2 | XP_006250090.1 | 8 - 9 | 1230 | 84 | ||
| GenBank mRNA | BC086987.1 | - | 8 - 9 | 1099 | 7129718 | 84 |
| U19614.1 | - | 7 - 8 | 888 | 84 | ||
| U20286.1 | - | 8 - 9 | 556 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P059507 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P103670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116778 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P103670 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P059507 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P103670 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P059507 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P059507 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P103670 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P582144 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701885330 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15517 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79996 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701885330 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10768921 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768920 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768925 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768916 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768929 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768927 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768917 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768913 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768915 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768926 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768923 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768914 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768928 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768930 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768922 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768919 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768918 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10768912 | - | - |
| Rat Genome 230 2.0 Array | 1388024_s_at | - | - |
| Rat Genome 230 2.0 Array | 1387917_at | - | - |
| Rat Genome 230 2.0 Array | 1388024_PM_s_at | - | - |
| Rat Genome 230 2.0 Array | 1387917_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387917_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387917_at | - | - |
| Affymetrix HT Rat Focus Array | 1388024_PM_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1388024_s_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300011745.rn.1 | TC1300001689.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300011736.rn.1 | TC1300001689.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300011742.rn.1 | TC1300001689.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300011738.rn.1 | TC1300001689.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300011741.rn.1 | TC1300001689.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300011740.rn.1 | TC1300001689.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387917_at | - | - |
| Rat Expression Set 230 | 1388024_s_at | - | - |
| Rat Expression Set 230 | 1388024_PM_s_at | - | - |
| Rat Expression Set 230 | 1387917_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768912 | - | - |
| Rat Genome U34 Set | U19614_g_at | - | - |
| Rat Genome U34 Set | U19614_at | - | - |
| Rat Genome U34 Set | rc_AI071690_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22150538 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350654 | - | - |
| RatRef-12 Expression BeadChip | ILMN_55236 | - | - |

Tor1aip1
Genomic Map