Assay Details
| Gene Symbol: Rab7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133590.1 | NP_598274.1 | 4 - 5 | 839 | 112 | |
| XM_006249744.2 | XP_006249806.1 | 5 - 6 | 816 | 112 | ||
| XM_006249745.2 | XP_006249807.1 | 5 - 6 | 673 | 112 | ||
| XM_008769422.1 | XP_008767644.1 | 3 - 4 | 369 | 112 | ||
| GenBank mRNA | FQ233577.1 | - | 4 - 5 | 605 | 112 | |
| FQ234855.1 | - | 4 - 5 | 605 | 112 | ||
| X96663.1 | - | 4 - 5 | 839 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab7l1
Genomic Map