Assay Details
| Gene Symbol: Mall | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014182.1 | NP_001014204.1 | 2 - 3 | 305 | 78 | |
| XM_008762181.1 | XP_008760403.1 | 2 - 3 | 342 | 78 | ||
| GenBank mRNA | BC090079.1 | - | 2 - 3 | 305 | 7126923 | 78 |
More Information
Updated Mapping Information
Set Membership: |
|

Mall
Genomic Map