Assay Details
| Gene Symbol: LOC103690017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234342.2 | XP_006234404.2 | 7 - 8 | 517 | 113 | |
| XM_008761952.1 | XP_008760174.1 | 7 - 8 | 1967 | 113 | ||
| Gene Symbol: Tlk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107734.1 | NP_001101204.1 | 684 | 113 | ||
| XM_008761896.1 | XP_008760118.1 | 7 - 8 | 735 | 113 | ||
| XM_008761897.1 | XP_008760119.1 | 1967 | 113 | |||
| XM_008761898.1 | XP_008760120.1 | 7 - 8 | 896 | 113 | ||
| XM_008761899.1 | XP_008760121.1 | 517 | 113 | |||
More Information
Updated Mapping Information
Set Membership: |
|

LOC103690017
Genomic Map