Assay Details
| Gene Symbol: Secisbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168527.1 | NP_001161999.1 | 15 - 16 | 2492 | 71 | |
| XM_008762152.1 | XP_008760374.1 | 15 - 16 | 2461 | 71 | ||
| GenBank mRNA | FQ213552.1 | - | 2 - 3 | 257 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Secisbp2l
Genomic Map