Assay Details
| Gene Symbol: Xirp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201989.1 | NP_973718.1 | 3 - 4 | 424 | 132 | |
| GenBank mRNA | AF466694.1 | - | 3 - 4 | 424 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xirp2
Genomic Map