Assay Details
| Gene Symbol: Fahd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134834.1 | NP_001128306.1 | 5 - 6 | 838 | 70 | |
| XM_006234960.2 | XP_006235022.1 | 5 - 6 | 856 | 70 | ||
| GenBank mRNA | BC166933.1 | - | 5 - 6 | 838 | 6921785 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Fahd2a
Genomic Map