Assay Details
| Gene Symbol: Arhgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107747.2 | NP_001101217.2 | 4 - 5 | 450 | 147 | |
| XM_006234551.1 | XP_006234613.1 | 5 - 6 | 487 | 147 | ||
| XM_006234552.2 | XP_006234614.1 | 5 - 6 | 517 | 147 | ||
| XM_006234553.2 | XP_006234615.1 | 5 - 6 | 487 | 147 | ||
| XM_006234554.1 | XP_006234616.1 | 6 - 7 | 529 | 147 | ||
| XM_006234556.1 | XP_006234618.1 | 5 - 6 | 464 | 147 | ||
| XM_006234557.2 | XP_006234619.1 | 5 - 6 | 494 | 147 | ||
| XM_006234558.2 | XP_006234620.1 | 4 - 5 | 435 | 147 | ||
| XM_006234559.2 | XP_006234621.1 | 2 - 3 | 1873 | 147 | ||
| XM_006234560.2 | XP_006234622.1 | 4 - 5 | 476 | 147 | ||
| GenBank mRNA | CK480577.1 | - | 4 - 5 | 450 | 147 | |
| FQ224002.1 | - | 4 - 5 | 410 | 147 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap1
Genomic Map