Assay Details
| Gene Symbol: Sema6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107768.1 | NP_001101238.1 | 2 - 3 | 601 | 66 | |
| XM_008762171.1 | XP_008760393.1 | 1 - 2 | 3974 | 66 | ||
| XM_008762172.1 | XP_008760394.1 | 1 - 2 | 3974 | 66 | ||
| XM_008762173.1 | XP_008760395.1 | 1 - 2 | 3974 | 66 | ||
| XM_008762174.1 | XP_008760396.1 | 1 - 2 | 3974 | 66 | ||
| XM_008762175.1 | XP_008760397.1 | 1 - 2 | 3974 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema6d
Genomic Map