Assay Details
| Gene Symbol: Knstrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004264.1 | NP_001004264.1 | 7 - 8 | 802 | 107 | |
| XM_006234757.2 | XP_006234819.1 | 7 - 8 | 838 | 107 | ||
| XM_006234758.2 | XP_006234820.1 | 7 - 8 | 659 | 107 | ||
| GenBank mRNA | BC079438.1 | - | 7 - 8 | 802 | 7115344 | 107 |
More Information
Updated Mapping Information
Set Membership: |
|

Knstrn
Genomic Map