Assay Details
| Gene Symbol: Prg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106487.1 | NP_001099957.1 | 4 - 5 | 690 | 63 | |
| XM_006234449.1 | XP_006234511.1 | 5 - 6 | 726 | 63 | ||
| XM_006234450.1 | XP_006234512.1 | 5 - 6 | 709 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prg3
Genomic Map