Assay Details
| Gene Symbol: Epb42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108590.2 | NP_001102060.1 | 3 - 4 | 547 | 77 | |
| GenBank mRNA | BC168935.1 | - | 3 - 4 | 547 | 9101840 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|

Epb42
Genomic Map