Assay Details
| Gene Symbol: Zfp106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762134.1 | XP_008760356.1 | 1 - 2 | 149 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008762134.1 transcripts.
Set Membership: |
|

Zfp106
Genomic Map