Assay Details
| Gene Symbol: Nup62cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234254.2 | XP_006234316.1 | 7 - 8 | 914 | 60 | |
| XM_006234255.2 | XP_006234317.1 | 7 - 8 | 808 | 60 | ||
| XM_006234257.2 | XP_006234319.1 | 7 - 8 | 943 | 60 | ||
| XM_008773468.1 | XP_008771690.1 | 7 - 8 | 862 | 60 | ||
| XM_008773470.1 | XP_008771692.1 | 7 - 8 | 779 | 60 | ||
| GenBank mRNA | BC160918.1 | - | 7 - 8 | 888 | 60 | |
More Information
Set Membership: |
|

Nup62cl
Genomic Map