Assay Details
| Gene Symbol: Tcp11l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109202.1 | NP_001102672.1 | 9 - 10 | 1540 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P053832 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P053827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122217 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053832 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053827 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053832 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053827 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P053827 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P053832 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P206215 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76176 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10847930 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847929 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847928 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847925 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847926 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847931 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847927 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10847924 | - | - |
| Rat Genome 230 2.0 Array | 1397737_at | - | - |
| Rat Genome 230 2.0 Array | 1397737_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300028886.rn.1 | TC0300003791.rn.1 | RTA TC |
| Rat Expression Set 230 | 1397737_PM_at | - | - |
| Rat Expression Set 230 | 1397737_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847924 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20752504 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352865 | - | - |

Tcp11l1
Genomic Map