Assay Details
| Gene Symbol: Sqrdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047913.2 | NP_001041378.2 | 1 - 2 | 268 | 79 | |
| XM_006234887.2 | XP_006234949.1 | 2 - 3 | 340 | 79 | ||
| GenBank mRNA | AY325246.1 | - | 2 - 3 | 102 | 79 | |
| BC107459.1 | - | 2 - 3 | 276 | 79 | ||
| BC158559.1 | - | 1 - 2 | 268 | 7456444 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sqrdl
Genomic Map