Assay Details
| Gene Symbol: Sqrdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047913.2 | NP_001041378.2 | 5 - 6 | 899 | 98 | |
| XM_006234887.2 | XP_006234949.1 | 6 - 7 | 971 | 98 | ||
| GenBank mRNA | BC158559.1 | - | 5 - 6 | 899 | 7456444 | 98 |
More Information
Updated Mapping Information
Set Membership: |
|

Sqrdl
Genomic Map