Assay Details
| Gene Symbol: Ext2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107751.1 | NP_001101221.1 | 7 - 8 | 1372 | 61 | |
| XM_006234576.2 | XP_006234638.2 | 7 - 8 | 1401 | 61 | ||
| GenBank mRNA | BC166748.1 | - | 7 - 8 | 1359 | 5599208 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|

Ext2
Genomic Map