Assay Details
| Gene Symbol: Rmdn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014046.2 | NP_001014068.1 | 2 - 3 | 281 | 78 | |
| GenBank mRNA | BC082081.1 | - | 2 - 3 | 243 | 7133344 | 78 |
| DN931790.1 | - | 2 - 3 | 313 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rmdn3
Genomic Map