Assay Details
| Gene Symbol: Blvra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053850.1 | NP_446302.1 | 4 - 5 | 452 | 98 | |
| XM_006234894.2 | XP_006234956.1 | 5 - 6 | 592 | 98 | ||
| XM_006234895.2 | XP_006234957.1 | 5 - 6 | 577 | 98 | ||
| GenBank mRNA | BC078766.1 | - | 4 - 5 | 464 | 7126157 | 98 |
| FQ213399.1 | - | 5 - 6 | 531 | 98 | ||
| FQ231949.1 | - | 5 - 6 | 533 | 98 | ||
| M81681.1 | - | 4 - 5 | 452 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Blvra
Genomic Map