Assay Details
| Gene Symbol: Zfp846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134585.2 | NP_001128057.1 | 1 - 2 | 277 | 93 | |
| XM_008765959.1 | XP_008764181.1 | 1 - 2 | 512 | 93 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001134585.2,XM_008765959.1 transcripts.
Set Membership: |
|

Zfp846
Genomic Map