Assay Details
| Gene Symbol: Casp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130422.1 | NP_569106.1 | 2 - 3 | 337 | 140 | |
| XM_008765891.1 | XP_008764113.1 | 3 - 4 | 140 | |||
| GenBank mRNA | AF317633.1 | - | 2 - 3 | 337 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casp12
Genomic Map