Assay Details
| Gene Symbol: Mrpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108754.1 | NP_001102224.1 | 4 - 5 | 525 | 56 | |
| GenBank mRNA | FQ215152.1 | - | 4 - 5 | 391 | 56 | |
| FQ226445.1 | - | 4 - 5 | 391 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl4
Genomic Map