Assay Details
| Gene Symbol: Prtfdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106127.1 | NP_001099597.1 | 8 - 9 | 669 | 77 | |
| XM_008771860.1 | XP_008770082.1 | 9 - 10 | 791 | 77 | ||
| GenBank mRNA | BC166887.1 | - | 8 - 9 | 666 | 6890546 | 77 |
| BC166923.1 | - | 8 - 9 | 666 | 6919422 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P220256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170450 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P220256 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P220256 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P220256 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P220256 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74376 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49928 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9122 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102031 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78146 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15124 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10796695 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799970 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799976 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799973 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799974 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799972 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799969 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799975 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799971 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10796694 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10799968 | - | - |
| Rat Genome 230 2.0 Array | 1374784_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374784_at | - | - |
| Affymetrix HT Rat Focus Array | 1374784_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374784_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700015668.rn.1 | TC1700002346.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700015670.rn.1 | TC1700002346.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374784_PM_at | - | - |
| Rat Expression Set 230 | 1374784_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799968 | - | - |
| Rat Genome U34 Set | rc_AA799402_at | - | - |
| Rat Genome U34 Set | rc_AI104491_s_at | - | - |
| Rat Genome U34 Set | rc_AI146137_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21668019 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370958 | - | - |

Prtfdc1
Genomic Map