Assay Details
| Gene Symbol: Olah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022705.1 | NP_073196.1 | 1 - 2 | 107 | 137 | |
| XM_008771886.1 | XP_008770108.1 | 2 - 3 | 260 | 137 | ||
| GenBank mRNA | M16200.1 | - | 1 - 2 | 107 | 137 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the M16200.1,NM_022705.1,XM_008771886.1 transcripts.
Set Membership: |
|

Olah
Genomic Map