Assay Details
| Gene Symbol: Taf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271342.1 | NP_001258271.1 | 4 - 5 | 2457 | 92 | |
| XM_006254228.2 | XP_006254290.1 | 4 - 5 | 2457 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf3
Genomic Map