Assay Details
| Gene Symbol: Sephs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104630.1 | NP_001098100.1 | 8 - 9 | 1330 | 74 | |
| XM_006254266.2 | XP_006254328.1 | 8 - 9 | 2957 | 74 | ||
| GenBank mRNA | AY724491.1 | - | 8 - 9 | 1330 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P823025 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P148776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87551 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P823025 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P148776 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P823025 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P148776 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P148776 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P823025 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P765356 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1035807 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100123 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92520 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10799569 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799574 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799572 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799575 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799573 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799570 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799577 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799576 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799571 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10799568 | - | - |
| Rat Genome 230 2.0 Array | 1385610_at | - | - |
| Rat Genome 230 2.0 Array | 1380362_at | - | - |
| Rat Genome 230 2.0 Array | 1393134_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393256_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385610_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389215_at | - | - |
| Rat Genome 230 2.0 Array | 1389215_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393256_at | - | - |
| Rat Genome 230 2.0 Array | 1381650_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381650_at | - | - |
| Rat Genome 230 2.0 Array | 1393134_at | - | - |
| Rat Genome 230 2.0 Array | 1380362_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393256_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393256_at | - | - |
| Affymetrix HT Rat Focus Array | 1389215_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385610_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381650_at | - | - |
| Affymetrix HT Rat Focus Array | 1385610_at | - | - |
| Affymetrix HT Rat Focus Array | 1380362_at | - | - |
| Affymetrix HT Rat Focus Array | 1380362_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389215_at | - | - |
| Affymetrix HT Rat Focus Array | 1381650_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700014774.rn.1 | TC1700002212.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700014771.rn.1 | TC1700002212.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700014773.rn.1 | TC1700002212.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700014772.rn.1 | TC1700002212.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389215_PM_at | - | - |
| Rat Expression Set 230 | 1389215_at | - | - |
| Rat Expression Set 230 | 1393134_at | - | - |
| Rat Expression Set 230 | 1381650_PM_at | - | - |
| Rat Expression Set 230 | 1380362_at | - | - |
| Rat Expression Set 230 | 1385610_PM_at | - | - |
| Rat Expression Set 230 | 1393256_PM_at | - | - |
| Rat Expression Set 230 | 1393256_at | - | - |
| Rat Expression Set 230 | 1381650_at | - | - |
| Rat Expression Set 230 | 1393134_PM_at | - | - |
| Rat Expression Set 230 | 1385610_at | - | - |
| Rat Expression Set 230 | 1380362_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799568 | - | - |
| Rat Genome U34 Set | rc_AA998097_at | - | - |
| Rat Genome U34 Set | rc_AI070739_at | - | - |
| Rat Genome U34 Set | AA801070_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22168787 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1649947 | - | - |

Sephs1
Genomic Map