Assay Details
| Gene Symbol: Ccdc110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771274.1 | XP_008769496.1 | 79 | 86 | ||
| GenBank mRNA | BC166517.1 | - | 79 | 7132003 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166517.1, XM_008771274.1 transcripts.
Set Membership: |
|

Ccdc110
Genomic Map