Assay Details
| Gene Symbol: Ccdc110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771274.1 | XP_008769496.1 | 287 | 112 | ||
| GenBank mRNA | BC166517.1 | - | 287 | 7132003 | 112 | |
| Gene Symbol: LOC100911140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771277.1 | XP_008769499.1 | 2 - 3 | 205 | 112 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC166517.1,XM_008771274.1 transcripts.
Set Membership: |
|

Ccdc110
Genomic Map