Assay Details
| Gene Symbol: Rwdd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034994.1 | NP_001030166.1 | 1 - 2 | 229 | 125 | |
| GenBank mRNA | BC092581.1 | - | 1 - 2 | 229 | 7369199 | 125 |
| FQ229862.1 | - | 1 - 2 | 270 | 125 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC092581.1,NM_001034994.1 transcripts.
Set Membership: |
|

Rwdd4
Genomic Map