Assay Details
| Gene Symbol: Rwdd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034994.1 | NP_001030166.1 | 2 - 3 | 316 | 157 | |
| GenBank mRNA | BC092581.1 | - | 2 - 3 | 316 | 7369199 | 157 |
| FQ229862.1 | - | 2 - 3 | 357 | 157 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rwdd4
Genomic Map