Assay Details
| Gene Symbol: Pdgfrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011921.1 | NP_001011921.1 | 4 - 5 | 880 | 81 | |
| GenBank mRNA | BC086555.1 | - | 4 - 5 | 880 | 7305039 | 81 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P160403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12215 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P160403 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P160403 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P160403 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P365794 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P299835 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8551 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77968 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28628 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38424 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56923 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10788398 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791899 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791895 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791898 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791894 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791896 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791897 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10791893 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10788397 | - | - |
| Rat Genome 230 2.0 Array | 1374616_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372457_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374616_at | - | - |
| Rat Genome 230 2.0 Array | 1372457_at | - | - |
| Affymetrix HT Rat Focus Array | 1374616_at | - | - |
| Affymetrix HT Rat Focus Array | 1374616_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600012729.rn.1 | TC1600001704.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600012730.rn.1 | TC1600001704.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374616_at | - | - |
| Rat Expression Set 230 | 1374616_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791893 | - | - |
| Rat Genome U34 Set | rc_AI177015_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21862360 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376295 | - | - |

Pdgfrl
Genomic Map