Assay Details
| Gene Symbol: Cnot7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107313.1 | NP_001100783.1 | 3 - 4 | 606 | 100 | |
| XM_008771290.1 | XP_008769512.1 | 3 - 4 | 779 | 100 | ||
| XM_008771291.1 | XP_008769513.1 | 3 - 4 | 749 | 100 | ||
| XM_008771292.1 | XP_008769514.1 | 3 - 4 | 649 | 100 | ||
| GenBank mRNA | BC167766.1 | - | 3 - 4 | 565 | 7455944 | 100 |
More Information
Updated Mapping Information
Set Membership: |
|

Cnot7
Genomic Map