Assay Details
| Gene Symbol: Tlr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198791.1 | NP_942086.1 | 2 - 3 | 108 | ||
| GenBank mRNA | AB116229.1 | - | 2 - 3 | 108 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB116229.1,NM_198791.1 transcripts.
Set Membership: |
|

Tlr3
Genomic Map