Assay Details
| Gene Symbol: Zmym5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728259.4 | XP_002728305.2 | 8 | 70 | ||
| XM_006252082.2 | XP_006252144.1 | 7 | 70 | |||
| XM_006252083.2 | XP_006252145.1 | 7 | 70 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zmym5
Genomic Map