Assay Details
| Gene Symbol: Pabpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135008.1 | NP_001128480.1 | 1 - 2 | 355 | 82 | |
| XM_008770659.1 | XP_008768881.1 | 2 - 3 | 2010 | 82 | ||
| GenBank mRNA | U94858.1 | - | 1 - 2 | 58 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pabpn1
Genomic Map