Assay Details
Gene Symbol: Lats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107267.1 | NP_001100737.1 | 6 - 7 | 2809 | 80 | |
XM_008770748.1 | XP_008768970.1 | 7 - 8 | 2854 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
protein phosphorylation
mitotic nuclear division
hormone-mediated signaling pathway
keratinocyte differentiation
cellular protein localization
hippo signaling
intracellular signal transduction
negative regulation of cyclin-dependent protein serine/threonine kinase activity
regulation of organ growth
negative regulation of canonical Wnt signaling pathway
protein serine/threonine kinase activity
ATP binding
G1/S transition of mitotic cell cycle
protein phosphorylation
mitotic nuclear division
hormone-mediated signaling pathway
keratinocyte differentiation
cellular protein localization
hippo signaling
intracellular signal transduction
negative regulation of cyclin-dependent protein serine/threonine kinase activity
regulation of organ growth
negative regulation of canonical Wnt signaling pathway
protein serine/threonine kinase activity
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P762252 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P012427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82772 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P762252 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012427 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P762252 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012427 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P762252 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P012427 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P20430 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2529 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17720 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10784165 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784166 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784169 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784168 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784170 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784171 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784167 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10784164 | - | - |
Rat Genome 230 2.0 Array | 1390207_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390207_at | - | - |
Affymetrix HT Rat Focus Array | 1390207_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390207_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1500011850.rn.1 | TC1500001783.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500011851.rn.1 | TC1500001783.rn.1 | RTA TC |
Rat Expression Set 230 | 1390207_at | - | - |
Rat Expression Set 230 | 1390207_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784164 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21964570 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373182 | - | - |
