Assay Details
| Gene Symbol: Micu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134396.2 | NP_599223.2 | 9 - 10 | 933 | 121 | |
| GenBank mRNA | DY317464.1 | - | 7 - 8 | 583 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P147159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51866 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P147160 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P147160 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P147160 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13259 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P151751 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85586 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10784209 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784202 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784207 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784203 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784205 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784210 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784206 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784211 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784200 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784204 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784201 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784208 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10784199 | - | - |
| Rat Genome 230 2.0 Array | 1372105_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369419_at | - | - |
| Rat Genome 230 2.0 Array | 1372105_at | - | - |
| Rat Genome 230 2.0 Array | 1369419_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369419_at | - | - |
| Affymetrix HT Rat Focus Array | 1372105_at | - | - |
| Affymetrix HT Rat Focus Array | 1369419_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372105_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500011929.rn.1 | TC1500001789.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500011931.rn.1 | TC1500001789.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500011928.rn.1 | TC1500001789.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500011932.rn.1 | TC1500001789.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500011930.rn.1 | TC1500001789.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369419_at | - | - |
| Rat Expression Set 230 | 1372105_PM_at | - | - |
| Rat Expression Set 230 | 1372105_at | - | - |
| Rat Expression Set 230 | 1369419_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784199 | - | - |
| Rat Genome U34 Set | rc_AA899956_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21175511 | - | - |

Micu2
Genomic Map