Assay Details
| Gene Symbol: Bcl2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021850.2 | NP_068622.1 | 1 - 2 | 103 | 99 | |
| XM_008770692.1 | XP_008768914.1 | 1 - 2 | 111 | 99 | ||
| GenBank mRNA | AY185098.1 | - | 1 - 2 | 74 | 99 | |
| BC074021.1 | - | 1 - 2 | 103 | 7105621 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY185098.1, BC074021.1, NM_021850.2, XM_008770692.1 transcripts.
Set Membership: |
|

Bcl2l2
Genomic Map