Assay Details
| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105727.2 | NP_001099197.2 | 1 - 2 | 207 | 68 | |
| GenBank mRNA | CB783917.1 | - | 1 - 2 | 207 | 68 | |
| D45247.1 | - | 1 - 2 | 205 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the D45247.1 transcripts.
Set Membership: |
|

Psmb5
Genomic Map