Assay Details
| Gene Symbol: Mrpl52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108375.2 | NP_001101845.2 | 3 - 4 | 179 | 111 | |
| XM_006251976.2 | XP_006252038.1 | 3 - 4 | 204 | 111 | ||
| GenBank mRNA | BC166922.1 | - | 3 - 4 | 179 | 6919316 | 111 |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl52
Genomic Map