Assay Details
| Gene Symbol: Rcbtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108380.1 | NP_001101850.1 | 7 - 8 | 1225 | 65 | |
| XM_008770759.1 | XP_008768981.1 | 8 - 9 | 1051 | 65 | ||
| GenBank mRNA | AY724518.1 | - | 6 - 7 | 871 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rcbtb1
Genomic Map